missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLIT3 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547519
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.20 EU per 1 μg of the antibody by the LAL method.
SLIT3 Polyclonal antibody specifically detects SLIT3 in Human, Mouse, Rat samples. It is validated for Neutralization, Western Blot.Specifications
SLIT3 | |
Polyclonal | |
Unconjugated | |
SLIT3 | |
b2b2362.1Clo; KIAA0814; Megf5; Multiple EGF-like domains protein 5; multiple epidermal growth factor-like domains 5; multiple epidermal growth factor-like domains protein 5; SLIL2; SLIL2FLJ10764; slit guidance ligand 3; slit homolog 3; slit homolog 3 (Drosophila); slit homolog 3 protein; SLIT1; slit2; SLIT3; Slit-3; UNQ691/PRO1336 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
20564, 6586, 83467 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O75094, O88280, Q9WVB4 | |
SLIT3 | |
Mouse myeloma cell line NS0-derived recombinant mouse Slit3 Ser27-His901 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |