Learn More
Sleeping Beauty Transposase Goat anti-Multi-species, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2798
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13467794
  • £446.88 / 100µg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Sleeping Beauty Transposase Polyclonal antibody AF2798 Specifically detects Sleeping Beauty Transposase in Many Species samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
Sleeping Beauty Transposase
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Affinity Purified
E. coli-derived recombinant Sleeping Beauty Transposase
Immunogen affinity purified
RUO
Primary
Detects Sleeping Beauty Transposase in direct ELISAs and Western blots.
Multi-Species
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Reconstitute at 0.2 mg/mL in sterile PBS.