missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGLT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534208
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon (100%) Monkey (86%).
SGLT1 Polyclonal antibody specifically detects SGLT1 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
SGLT1 | |
Polyclonal | |
Unconjugated | |
Slc5a1 | |
D22S675; High affinity sodium-glucose cotransporter; HSGLT1; MGC93553; Na; Na(+)/glucose cotransporter 1; Na+/glucose cotransporter 1; NAGT; Sglt1; SLC5A1; sodium glucose cotransporter 1; sodium/glucose cotransporter 1; sodium-glucose cotransporter 1; sodium-glucose cotransporter-like 1; solute carrier family 5 (sodium/glucose cotransporter), member 1; solute carrier family 5 member 1; solute carrier family 5, member 1; solute carrier family 5, member alpha 1; Solute carrier family 5, member alpha 1 (Na+/glucose cotransporter) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6523 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P13866 | |
Slc5a1 | |
Synthetic 14 amino acid peptide from internal region of human SLC5A1 / SGLT1. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |