Learn More
SLC44A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534205
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla (100%); Gibbon, Monkey, Marmoset (94%); Orangutan, Galago, Dog (88%); Rat, Hamster, Elephant, Panda, Horse, Pig, Guinea pig (81%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
SLC44A2 Polyclonal antibody specifically detects SLC44A2 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
SLC44A2 | |
Polyclonal | |
Unconjugated | |
SLC44A2 | |
1110028E10Rik; Choline transporter-like protein 2; Ctl2; IESCA; inner ear supporting cell antigen; PP1292; PSEC0210; RGD1309680; SLC44A2; solute carrier family 44 (choline transporter), member 2; solute carrier family 44 member 2; solute carrier family 44, member 2; solute carrier family 44, member 2 tv2; testicular tissue protein Li 47 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57153 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q8IWA5 | |
SLC44A2 | |
Synthetic 16 amino acid peptide from internal region of human SLC44A2. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |