Learn More
SLC38A5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542504
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: GIRAYEQLGQ RAFGPAGKVV VATVICLHNV GAMSSYLFII KSELPLVIGT Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 85%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 100%
SLC38A5 Polyclonal antibody specifically detects SLC38A5 in Human samples. It is validated for Western Blot.Specifications
SLC38A5 | |
Polyclonal | |
Unconjugated | |
SLC38A5 | |
amino acid transport system N2; amino acid transporter system N2; C81234; E330031E14; JM24; PP7194; SLC38A5; Sn2; Snat5; sodium-coupled neutral amino acid transporter 5; sodium-coupled neutral amino acid transporter 5; LOW QUALITY PROTEIN: sodium-coupled neutral amino acid transporter 5; solute carrier family 38 (amino acid transporter), member 5; solute carrier family 38 member 5; solute carrier family 38, member 5; system N transporter 2; transport system N, protein 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
92745 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8WUX1 | |
SLC38A5 | |
Synthetic peptide directed towards the N-terminal of human SLC38A5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |