missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC38A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542419
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: LLKNLGYLGY TSGFSLSCMV FFLIVVIYKK FQIPCIVPEL NSTISANSTN Sequence homology: Cow: 85%; Dog: 91%; Guinea Pig: 92%; Horse: 86%; Human: 100%; Rat: 92%
SLC38A1 Polyclonal antibody specifically detects SLC38A1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
SLC38A1 | |
Polyclonal | |
Unconjugated | |
SLC38A1 | |
AA408026; AA409865; AL022800; Amino acid transporter A1; amino acid transporter system A1; ATA1; AU015942; GlnT; glutamine transporter; MNat2; Nat2; N-system amino acid transporter 2; rATA1; S38A1; Sa2; SAT1; SLC38A1; SNAT 1; Snat1; sodium-coupled neutral amino acid transporter 1; Solute carrier family 38 member 1; solute carrier family 38, member 1; system A amino acid transporter 1; System A transporter 2; system N amino acid transporter 1 | |
Rabbit | |
Protein A | |
RUO | |
81539 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H2H9 | |
SLC38A1 | |
Synthetic peptide directed towards the middle region of human SLC38A1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |