Learn More
SLC35C1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542475
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: TSISMVFLNK YLLDSPSLRL DTPIFVTFYQ CLVTTLLCKG LSALAACCPG Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 92%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 100%; Rat: 93%
SLC35C1 Polyclonal antibody specifically detects SLC35C1 in Human samples. It is validated for Western Blot.Specifications
SLC35C1 | |
Polyclonal | |
Unconjugated | |
SLC35C1 | |
CDG2C; E430007K15Rik; fuct1; GDP-fucose transporter; GDP-fucose transporter 1; GDP-Fuc-Tr; golgi GDP-fucose translocator; multispan transmembrane protein fuct1; putative golgi GDP-fucose translocator; SLC35C1; solute carrier family 35 (GDP-fucose transporter), member C1; solute carrier family 35 member C1; solute carrier family 35, member C1 | |
Rabbit | |
Protein A | |
RUO | |
55343 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96A29 | |
SLC35C1 | |
Synthetic peptide directed towards the N-terminal of human SLC35C1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |