missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC26A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542487
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: LYSLTGLDAG CMAARRKEGG SETGVGEGGP AQGEDLGPVS TRAALVPAAA Sequence homology: Human: 100%
SLC26A1 Polyclonal antibody specifically detects SLC26A1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
SLC26A1 | |
Polyclonal | |
Unconjugated | |
SLC26A1 | |
CAON; EDM4; SAT1; SAT-1; SLC26A1; solute carrier family 26 (anion exchanger), member 1; solute carrier family 26 (sulfate transporter), member 1; solute carrier family 26 member 1; solute carrier family 26, member 1; sulfate anion tranporter AT1; sulfate anion transporter 1; sulfate anion transporter-1; sulfate/anion transporter SAT-1 protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10861 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H2B4 | |
SLC26A1 | |
Synthetic peptide directed towards the C-terminal of human SLC26A1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |