Learn More
SLC25A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542451
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: NKPMNPLITG VFGAIAGAAS VFGNTPLDVI KTRMQGLEAH KYRNTWDCGL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 100%; Zebrafish: 100%
SLC25A1 Polyclonal antibody specifically detects SLC25A1 in Human samples. It is validated for Western Blot.Specifications
SLC25A1 | |
Polyclonal | |
Unconjugated | |
SLC25A1 | |
1300019P08Rik; 2610100G11Rik; AI194714; Cic; Citrate transport protein; citrate transporter) member 1; clathrin heavy chain 2; clathrin heavy chain 2; uncharacterized protein LOC100053137; clathrin, heavy chain-like 1; CLTCL1; Ctp; D2L2AD; Dgsj; DiGeorge syndrome gene j; LOC100053137; mitochondrial tricarboxylate carrier; SEA; Slc20a3; SLC25A1; solute carrier family 20 (mitochondrial citrate transporter), member 3; solute carrier family 25 (mitochondrial carrier, citrate transporter), member 1; solute carrier family 25 (mitochondrial carrier; citrate transporter), member 1; solute carrier family 25 member 1; Tricarboxylate carrier protein; tricarboxylate transport protein, mitochondrial; txtp | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6576 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P53007 | |
SLC25A1 | |
Synthetic peptide directed towards the middle region of human SLC25A1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |