missing translation for 'onlineSavingsMsg'
Learn More

solute carrier family 22 (organic cation/ergothioneine transporter), member 4, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant SLC22A4.

Brand:  Abnova H00006583-A01.50uL

Additional Details : Weight : 0.00010kg

Product Code. 16165455

  • £189.00 / 50µL
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. The encoded protein is an organic cation transporter and plasma integral membrane protein containing eleven putative transmembrane domains as well as a nucleotide-binding site motif. Transport by this protein is at least partially ATP-dependent. [provided by RefSeq

Sequence: AGTPEHRCRVPDAANLSSAWRNNSVPLRLRDGREVPHSCSRYRLATIANFSALGLEPGRDVDLGQLEQESCLDGWEFSQDVYLSTVVTEWNLVCEDNWK
Specifications

Specifications

solute carrier family 22 (organic cation/ergothioneine transporter), member 4
Polyclonal
Mouse polyclonal antibody raised against a partial recombinant SLC22A4.
SLC22A4
MGC34546/MGC40524/OCTN1
Mouse
50 μL
Yes
6583
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
Unconjugated
50% glycerol
NM_003059
SLC22A4
SLC22A4 (NP_003050, 43 a.a. ∼ 141 a.a) partial recombinant protein with GST tag.
RUO
Primary
Human
Serum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers