missing translation for 'onlineSavingsMsg'
Learn More
Learn More
solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SLC13A2.
Brand: Abnova H00009058-A01.50uL
Additional Details : Weight : 0.00010kg
Description
Sequence: AMMVPIAHAVLDQLHSSQASSNVEEGSNNPTFELQEPSPQKEVTKLDNGQALPVTSASSEGRAHLSQKHLHLTQCSpecifications
solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant SLC13A2. | |
SLC13A2 | |
NADC1/NaDC-1 | |
Mouse | |
50 μL | |
Yes | |
9058 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_003984 | |
SLC13A2 | |
SLC13A2 (NP_003975, 146 a.a. ∼ 220 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |