Learn More
SIRT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA3200
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA3-200 detects human and rat SIRT2 and shows no cross-reactivity to other SIRT isoforms. PA3-200 has been successfully used in Western blot and immunocytochemistry procedures. By Western blot, this antibody detects an ∽48 kDa protein representing SIRT2 from SAOS2 cells. PA3-200 can be used for immunocytochemistry procedures. PA3-200 immunizing peptide corresponds to amino acid residues 341-352 from the human SIRT2 protein. This sequence is completely conserved between human and rat.
SIRT2 Polyclonal antibody specifically detects SIRT2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
SIRT2 | |
Polyclonal | |
Unconjugated | |
SIRT2 | |
5730427M03Rik; 5E5 antigen; mSIR2L2; NAD-dependent deacetylase sirtuin-2; NAD-dependent protein deacetylase sirtuin-2; Regulatory protein SIR2 homolog 2; silent information regulator 2; silent mating type information regulation 2, (S.cerevisiae, homolog)-like; sirtuin 2; SIR2; SIR2L; SIR2L2; SIR2-like protein 2; sir2-related protein type 2; Sirt2; sirtuin 2; sirtuin type 2; sirtuin-2 | |
Rabbit | |
NA | |
RUO | |
22933, 361532, 64383 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Conc. Not Determined | |
whole serum with 0.05% sodium azide | |
Q5RJQ4, Q8IXJ6, Q8VDQ8 | |
SIRT2 | |
Synthetic Peptide: D(341) E A R T T E R E K P Q(352) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |