Learn More
SIGNR1/CD209b Rat anti-Mouse, Clone: 258829, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1836
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13438965
  • £249.49 / 100µg
Estimated Shipment
28-06-2019
Description
Description
SIGNR1/CD209b Monoclonal antibody MAB1836 Specifically detects SIGNR1/CD209b in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
SIGNR1/CD209b
258829
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Cd209b
Mouse myeloma cell line NS0-derived recombinant mouse SIGNR1/CD209b Ser76-Gly325 Accession # Q8CJ91
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
170786
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q8CJ91
CD209 antigen like protein B, CD209b, DC SIGN related protein 1, DC SIGNR1, OtB7
Rat
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse SIGNR1/CD209b in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse SIGNR4 is observed.