Learn More
Siglec-F Rat anti-Mouse, Clone: 238023, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1706
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13438845
  • £337.93 / 500µg
Estimated Shipment date
from Supplier 27-09-2019
Description
Description
Siglec-F Monoclonal antibody MAB1706 Specifically detects Siglec-F in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Siglec-F
238023
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Siglecf
Mouse myeloma cell line NS0-derived recombinant mouse Siglec-F Asp18-Thr437 (predicted) Accession # Q920G3
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
27181
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q920G3
SAF2, sialic acid binding Ig like lectin 8, SIGLEC-8, SIGLEC8L, SiglecF
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Siglec-F in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Siglec-5, -7, -9, -10, or recombinant mouse Siglec-2, or -3 is observed.