Learn More
Siglec-7/CD328 Mouse anti-Human, Clone: 194212, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1138
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448385
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 26-02-2020
Description
Description
Siglec-7/CD328 Monoclonal antibody MAB1138 Specifically detects Siglec-7/CD328 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Siglec-7/CD328
194212
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SIGLEC7
Mouse myeloma cell line NS0-derived recombinant human Siglec-7/CD328 Gln19-Gly357 (predicted) Accession # Q9Y286
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
27036
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9Y286
Adhesion inhibitory receptor molecule 1, adhesion inhibitory receptor molecule 1, siglec-7, AIRM1, AIRM-1, AIRM1QA79 membrane protein, CD328, CD328 antigen, CDw328, D-siglec, p75, p75/AIRM1, QA79, sialic acid binding Ig-like lectin 7, sialic acid binding immunoglobulin-like lectin 7, sialic acid-binding Ig-like lectin 7, Siglec7, Siglec-7
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Siglec-7/CD328 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Siglec-2, -3, -5, or -9 is observed.