Learn More
Siglec-5/CD170 Mouse anti-Human, Clone: 194117, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1072
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13468305
  • £427.77 / 500µg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
Siglec-5/CD170 Monoclonal antibody MAB1072 Specifically detects Siglec-5/CD170 in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
Siglec-5/CD170
194117
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SIGLEC5
Mouse myeloma cell line NS0-derived recombinant human Siglec-5/CD170 Lys18-Thr434 Accession # O15389
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8778
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunohistochemistry
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
O15389
CD170, CD170 antigen, CD33 antigen-like 2, CD33L2, CD33L2OB binding protein-2, OBBP2, OB-BP2OB-binding protein 2, OBBP2sialic acid-binding Ig-like lectin 5, Obesity-binding protein 2, sialic acid binding Ig-like lectin 5, sialic acid-binding immunoglobulin-like lectin 5, Siglec5, SIGLEC-5
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Siglec-5/CD170 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Siglec-2, -3, -7, -9, or -14 is observed.