missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIGIRR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA520078
Code : LD
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is A549 cell lysate. PA5-20078 can be used with blocking peptide PEP-0196.
SIGIRR Polyclonal antibody specifically detects SIGIRR in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
SIGIRR | |
Polyclonal | |
Unconjugated | |
SIGIRR | |
AI256711; IL-1R8; MGC110992; Sigirr; single Ig and TIR domain containing; single Ig IL-1 receptor related protein; single Ig IL-1-related receptor; Single Ig IL-1R-related molecule; single Ig IL-1R-related protein; single immunoglobulin and toll-interleukin 1 receptor (TIR) domain; single immunoglobulin domain IL1R1 related; single immunoglobulin domain-containing IL1R-related protein; TIR8; Toll/interleukin-1 receptor 8; UNQ301/PRO342 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
24058, 59307 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q6IA17, Q9JLZ8 | |
SIGIRR | |
A synthetic peptide corresponding to 15 amino acids at the C-terminus of mouse SIGIRR. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |