missing translation for 'onlineSavingsMsg'
Learn More

SIGIRR Rat anti-Mouse, Clone: 161917, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1092

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458325

  • £353.77 / 500µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

SIGIRR Monoclonal antibody MAB1092 Specifically detects SIGIRR in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

SIGIRR
161917
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SIGIRR
Mouse myeloma cell line NS0-derived recombinant mouse SIGIRR, Met1-His117, Accession # Q9JLZ8
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
59307
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9JLZ8
single Ig IL-1-related receptor, Single Ig IL-1R-related molecule, single immunoglobulin and toll-interleukin 1 receptor (TIR) domain, single immunoglobulin domain IL1R1 related, Single immunoglobulin domain-containing IL1R-related protein, TIR8, TIR8MGC110992, Toll/interleukin-1 receptor 8
Rat
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects mouse SIGIRR in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant mouse (rm) IL-1 R1, rmIL-1 R2, rhIL-1 RAcP, recombinant human (rh) IL-1 R4, rmIL-18 R, recombinant rat IL-1 R6, rmIL-1 R7, rhIL-1 R8, rhIL-1 R9, or rhSIGIRR.
Mouse
Videos

For Research Use Only