Learn More
SHP-1 Rat anti-Human, Clone: 255402, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1878
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13458995
  • £271.93 / 100µg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
SHP-1 Monoclonal antibody MAB1878 Specifically detects SHP-1 in Human samples. It is validated for Immunocytochemistry
Specifications
Specifications
SHP-1/Nuclear Receptor SHP
255402
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human SHP-1.
Human
Immunocytochemistry
LYOPH
Immunocytochemistry 3-25 ug/mL
P29350
PTPN6
E. coli-derived recombinant human SHP-1 Ala205-Lys595 Accession # P29350
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8431
Reconstitute at 0.5 mg/mL in sterile PBS.