Learn More
F(ab')2-Donkey anti-Sheep IgG (H+L) Secondary Antibody, Invitrogen™
F(ab')2-Donkey Polyclonal Secondary Antibody
Brand: Invitrogen A24563
82.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Sheep IgG (H+L) Polyclonal antibody specifically detects Sheep IgG (H+L) in Ovine samples. It is validated for ELISA.Specifications
Sheep IgG (H+L) | |
Polyclonal | |
Unconjugated | |
Donkey | |
Affinity Chromatography | |
RUO | |
Ovine | |
Liquid |
ELISA | |
> 4.5 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.2 | |
Gamma Immunoglobins Heavy and Light chains | |
2 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |