missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHC Mouse anti-Human, Mouse, Rat, Clone: 47F4, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen LFMA0201
Additional Details : Weight : 0.99000kg
Description
A suggested positive control for this product is HeLa cells.
Signaling adapter that couples activated growth factor receptors to signaling pathways. Participates in a signaling cascade initiated by activated KIT and KITLG/SCF. Isoform p46Shc and isoform p52Shc, once phosphorylated, couple activated receptor tyrosine kinases to Ras via the recruitment of the GRB2/SOS complex and are implicated in the cytoplasmic propagation of mitogenic signals. Isoform p46Shc and isoform p52Shc may thus function as initiators of the Ras signaling cascade in various non-neuronal systems. Isoform p66Shc does not mediate Ras activation, but is involved in signal transduction pathways that regulate the cellular response to oxidative stress and life span. Isoform p66Shc acts as a downstream target of the tumor suppressor p53 and is indispensable for the ability of stress-activated p53 to induce elevation of intracellular oxidants, cytochrome c release and apoptosis. The expression of isoform p66Shc has been correlated with life span . Participates in signaling downstream of the angiopoietin receptor TEK/TIE2, and plays a role in the regulation of endothelial cell migration and sprouting angiogenesis.Specifications
SHC | |
Monoclonal | |
1 mg/mL | |
SHC1 | |
RP11-307C12.1, FLJ26504, SHC, SHCA | |
Mouse | |
Protein G | |
RUO | |
20416, 6464, 85385 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
47F4 | |
Unconjugated | |
P29353, P98083, Q5M824 | |
SHC1 | |
Recombit human protein purified from E.coli (His/ABD-Shc1) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |