Learn More
SGK2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533019
Code : PN
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey, Bovine (100%); Marmoset, Dog, Horse, Rabbit, Platypus (93%); Bat, Elephant, Panda, Opossum, Turkey, Chicken (87%); Xenopus (80%). For IHC(P), use proteinase K antigen retrieval. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
SGK2 Polyclonal antibody specifically detects SGK2 in Bovine, Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin).Specifications
SGK2 | |
Polyclonal | |
Unconjugated | |
SGK2 | |
AI098171; AW146006; dJ138B7.2; H-SGK2; Serine/threonine-protein kinase Sgk2; serum/glucocorticoid regulated kinase 2; serum/glucocorticoid-regulated kinase 2; Sgk2; SGK2, serine/threonine kinase 2; Sgkl | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10110, 517909 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9HBY8 | |
SGK2 | |
Synthetic 15 amino acid peptide from kinase domain of human SGK2. | |
50 μg | |
Primary | |
Bovine, Human | |
Antibody | |
IgG |