missing translation for 'onlineSavingsMsg'
Learn More

sFRP-3 Mouse anti-Human, Mouse, Clone: 133912, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB192-500

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449035

  • £390.72 / 500µg

Estimated Shipment date
from Supplier 09-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

sFRP-3 Monoclonal antibody MAB192500 Specifically detects sFRP-3 in Human, Mouse samples. It is validated for Western Blotting
Specifications

Specifications

sFRP-3/FRZB
133912
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
FRZB
Mouse myeloma cell line NS0-derived recombinant human sFRP-3 Ala33-Asn325 Accession # AAB51298
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2487
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
AAB51298
FIZ, FREOS1, Frezzled, Fritz, frizzled-related protein, Frizzled-related protein 1, FRP, FRP-3, FRZB, Frzb1, FrzB-1, FRZB1sFRP-3, FRZB-PEN, FZRB, hFIZ, secreted frizzled-related protein 3, sFRP3, SFRP3frezzled, SRFP3frizzled homolog-related
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human sFRP-3 in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant mouse (rm) sFRP-3 and no cross-reactivity with recombinant human (rh) sFRP-1, rmsFRP-2, or rhsFRP-4.
Human, Mouse
Videos
Documents