Learn More
sFRP-2 Rat anti-Mouse, Clone: 331022, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1169
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13468405
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 02-04-2020
Description
Description
sFRP-2 Monoclonal antibody MAB1169 Specifically detects sFRP-2 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
sFRP-2
331022
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SFRP2
Mouse myeloma cell line NS0-derived recombinant mouse sFRP-2 Leu25-Cys295 Accession # P97299
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
6423
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P97299
FRP2, FRP-2secreted apoptosis related protein 1, SARP-1, SARP1sFRP-2, SDF-5, Secreted apoptosis-related protein 1, secreted frizzled-related protein 2, sFRP2
Rat
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse sFRP-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) sFRP-1, recombinant mouse sFRP-3, or rhsFRP-4.