Learn More
SFMBT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543563
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: PGNCVLVLRE VLTLLINAAY KPSRVLRELQ LDKDSVWHGC GEVLKAKYKG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%
SFMBT1 Polyclonal antibody specifically detects SFMBT1 in Human samples. It is validated for Western Blot.Specifications
SFMBT1 | |
Polyclonal | |
Unconjugated | |
SFMBT1 | |
4930442N21Rik; 9330180L21Rik; AA536974; hSFMBT; Renal ubiquitous protein 1; RU1; Scm like with four mbt domains 1; Scm-like with four mbt domains 1; scm-like with four MBT domains protein 1; Scm-related gene containing four mbt domains; Scm-related gene containing four mbt domains 1; Scm-related gene product containing four mbt domains; SFMBT; SFMBT1; Smr; wu:fc12f05 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51460 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UHJ3 | |
SFMBT1 | |
Synthetic peptide directed towards the middle region of human SFMBT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |