Learn More
Human Serum Albumin Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Brand: Invitrogen PA172055
118.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-72055 detects Human serum albumin in human samples. PA1-72055 has been successfully used in ELISA and western blot procedures. The PA1-72055 immunogen is from purified normal human serum albumin.
Human Serum Albumin Polyclonal antibody specifically detects Human Serum Albumin in Human samples. It is validated for ELISA, Western Blot.Specifications
Human Serum Albumin | |
Polyclonal | |
Unconjugated | |
ALB | |
Alb; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; cell growth inhibiting protein 42; DKFZp779N1935; FDAH; GIG20; GIG42; growth-inhibiting protein 20; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341 | |
Chicken | |
IgY fraction | |
RUO | |
213 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.075% sodium azide; pH 7.2 | |
P02768 | |
ALB | |
Purified normal human serum albumin. | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgY |