Learn More
PAI1 Monoclonal Antibody (1D5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517171
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17171 targets SERPINE1 in FACS, IHC, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17171 immunogen is purified recombinant fragment of human SERPINE1 expressed in E. Coli. MA5-17171 detects SERPINE1 which has a predicted molecular weight of approximately 45kDa.
PAI1 Monoclonal antibody specifically detects PAI1 in Human, Mouse samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
PAI1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P05121, P22777 | |
Serpine1 | |
Purified recombinant fragment of human SERPINE1 expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
1D5 | |
Unconjugated | |
Serpine1 | |
Endothelial plasminogen activator inhibitor; nexin, plasminogen activator inhibitor type 1; PAI; PAI1; PAI-1; PAI1 protein; PAI1A; Pai1aa; Planh; Planh1; plas; plasminogen activator inhibitor; plasminogen activator inhibitor 1; plasminogen activator inhibitor I; plasminogen activator inhibitor, type I; plasminogen activator inhibitor-1; Plasminogen activator inhibitor-1 precursor (PAI-1) (Endothelial plasminogen activator inhibitor) (PAI); putative plasminogen activator inhibitor-1; RATPAI1A; serine (or cysteine) peptidase inhibitor, clade E, member 1; serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1; serine (or cysteine) proteinase inhibitor, clade E, member 1; serine (or cysteine) proteinase inhibitor, member 1; serpin E1; serpin family E member 1; serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1; SERPINE1; SERPINE-1 | |
Mouse | |
Protein G | |
RUO | |
18787, 5054 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |