Learn More
SERPINB13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA14286
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-4286 detects SERPINB13 from human samples. PA1-4286 has been successfully used in Western blot procedures. PA1-4286 immunizing peptide corresponds to amino acid residues 310-324 from human SERPINB13 protein.
SERPINB13 Polyclonal antibody specifically detects SERPINB13 in Human samples. It is validated for Western Blot.Specifications
SERPINB13 | |
Polyclonal | |
Unconjugated | |
SERPINB13 | |
5430417G24; HaCaT UV-repressible serpin; headpin; HSHUR7SEQ; HUR7; HURPIN; Peptidase inhibitor 13; PI13; PI-13; protease inhibitor 13 (hurpin, headpin); proteinase inhibitor 13; serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 13; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 13; serpin B13; serpin family B member 13; serpin peptidase inhibitor, clade B (ovalbumin), member 13; SERPINB13; UV-B repressed sequence, HUR 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5275 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9UIV8 | |
SERPINB13 | |
Synthetic Peptide: S(310) E H K A D Y S G M S S G S G(324) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |