Learn More
Serpin I2 Mouse anti-Human, Clone: 203402, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2235
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439285
  • £337.93 / 500µg
Estimated Shipment
27-06-2019
Description
Description
Serpin I2 Monoclonal antibody MAB2235 Specifically detects Serpin I2 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Serpin I2
203402
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Purified
SERPINI2
Mouse myeloma cell line NS0-derived recombinant human Serpin I2
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5276
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
MEPI, MEPIpancpin, Myoepithelium-derived serine protease inhibitor, Pancipin, Pancpin, Pancreas-specific protein TSA2004, Peptidase inhibitor 14, PI14, PI-14, PI14clade I (neuroserpin), member 2, protease inhibitor 14, serine (or cysteine) proteinase inhibitor, clade I (pancpin), member 2, serpin I2, serpin peptidase inhibitor, clade I (pancpin), member 2, SERPIN12
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Serpin I2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Serpin A1, A3, A4, A5, C1, F2, I1, recombinant mouse (rm) Serpin D1, or rmPEDF is observed.