Learn More
Serpin F2/alpha 2-Antiplasmin Mouse anti-Human, Clone: 236122, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1470
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13438665
  • £373.57 / 500µg
Estimated Shipment
09-07-2019
Description
Description
Serpin F2/alpha 2-Antiplasmin Monoclonal antibody MAB1470 Specifically detects Serpin F2/alpha 2-Antiplasmin in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Serpin F2/alpha 2-Antiplasmin
236122
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SERPINF2
Mouse myeloma cell line NS0-derived recombinant human Serpin F2/alpha 2 -Antiplasmin Met28-Lys491 Accession # P08697
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5345
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
P08697
AAPclade F (alpha-2 antiplasmin, alpha 2-Antiplasmin, Alpha-2-AP, Alpha-2-PI, Alpha-2-plasmin inhibitor, pigment epithelium derived factor), member 2, Serpin F2, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 2
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Serpin F2/alpha 2 -Antiplasmin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Serpin A1, A3, A4, A5, recombinant mouse Serpin C1, D1, F2, or recombinant mouse PEDF is observed.