Learn More
Serpin F1/PEDF Rat anti-Mouse, Clone: 199315, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1149
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13458395
  • £337.93 / 500µg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
Serpin F1/PEDF Monoclonal antibody MAB1149 Specifically detects Serpin F1/PEDF in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Serpin F1/PEDF
199315
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SERPINF1
E. coli-derived recombinant mouse Serpin F1/PEDF Asp43-Thr417 Accession # P97298
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5176
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P97298
Cell proliferation-inducing gene 35 protein, EPC-1, EPC-1PIG35, PEDF, PEDFpigment epithelium-derived factor, pigment epithelium derived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 1
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Serpin F1/PEDF in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human Serpin A1, A3, A5, C1, or F1.