Learn More
Serpin B5/Maspin Rat anti-Human, Clone: 305339, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2218
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439275
  • £249.49 / 100µg
Estimated Shipment
09-07-2019
Description
Description
Serpin B5/Maspin Monoclonal antibody MAB2218 Specifically detects Serpin B5/Maspin in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Serpin B5/Maspin
305339
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SERPINB5
E. coli-derived recombinant human Serpin B5 Met1-Pro375 Accession # P36952
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5268
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
P36952
maspin, Peptidase inhibitor 5, PI5, PI-5, PI5protease inhibitor 5 (maspin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 5, serpin B5, serpin peptidase inhibitor, clade B (ovalbumin), member 5
Rat
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Serpin B5/Maspin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Serpin A5, B6, B8 or B9 is observed.