missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A9/Centerin Mouse anti-Human, Clone: 585302, R&D Systems™
Mouse Monoclonal Antibody
Brand: RD Systems (Bio-Techne) MAB5429
Code : Z1
Additional Details : Weight : 0.00010kg
Description
Serpin A9/Centerin Monoclonal antibody MAB5429 Specifically detects Serpin A9/Centerin in Human samples. It is validated for ImmunohistochemistrySpecifications
Serpin A9/Centerin | |
585302 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Purified | |
Mouse | |
IgG2a | |
100 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Monoclonal | |
Detects human Serpin A9/Centerin in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Serpin A1, A3, A4, A5, A6, A11, A12, B6, B8, B9, C1, D1, E1, E2, F1, F2, G1, I1, I2, recombinant mouse (rm) SerpinA3N, and rmSerpinA9 is observed. | |
Human |
Immunohistochemistry | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
AAQ89063 | |
SERPINA9 | |
Mouse myeloma cell line NS0-derived recombinant human Serpin A9/Centerin Ala24-Thr417 Accession # AAQ89063 | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
327657 | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
For Research Use Only