missing translation for 'onlineSavingsMsg'
Learn More

Serpin A6 Rat anti-Mouse, Clone: 433903, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB4065

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460386

  • £249.49 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Serpin A6 Monoclonal antibody MAB4065 Specifically detects Serpin A6 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Serpin A6/Cortisol Binding Globulin
433903
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SERPINA6
Mouse myeloma cell line NS0-derived recombinant mouse Serpin A6, Val23-Ala397, Accession # Q06770
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
866
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q06770
antitrypsin), member 6, CBG, CBGcorticosteroid-binding globulin, corticosteroid binding globulin, member 6, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A6, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Transcortin
Rat
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects mouse Serpin A6 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse Serpin A1, A5, B8, C1, D1, E1, E2, F1, F2, I1, recombinant human Serpin A3, A4, B6, B9, G1, I2, or recombinant rat Serpin A6 is observed.
Mouse
Videos

For Research Use Only