Learn More
Serpin A3/alpha 1-Antichymotrypsin Mouse anti-Human, Clone: 213907, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1295
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448525
  • £390.72 / 500µg
Estimated Shipment date
from Supplier 06-07-2020
Description
Description
Serpin A3/alpha 1-Antichymotrypsin Monoclonal antibody MAB1295 Specifically detects Serpin A3/alpha 1-Antichymotrypsin in Human samples. It is validated for Western Blotting
Specifications
Specifications
Serpin A3/alpha 1-Antichymotrypsin
213907
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SERPINA3
Mouse myeloma cell line NS0-derived recombinant human Serpin A3 aa 26-423 Accession # P01011
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
12
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P01011
AACTMGC88254, ACTGIG25, alpha 1-Antichymotrypsin, alpha-1-antichymotrypsin, antitrypsin), member 3, Cell growth-inhibiting gene 24/25 protein, growth-inhibiting protein 24, growth-inhibiting protein 25, member 3, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 3, Serpin A3, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects recombinant human (rh)Serpin A3 in direct ELISAs and Western blots. In these formats, this antibody shows 5 - 25% cross-reactivity with rhSerpins A1 and A5 but no cross-reactivity with rhSerpins C1, D1, or F2.