missing translation for 'onlineSavingsMsg'
Learn More

SEPHS1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA534740

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15358611

  • £404.00 / 100µL
Estimated Shipment: 23-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Recommended positive controls: HepG2. Predicted reactivity: Mouse (100%), Rat (99%), Zebrafish (98%), Xenopus laevis (98%), Dog (100%), Chicken (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SEPHS1 Polyclonal antibody specifically detects SEPHS1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Specifications

Specifications

SEPHS1
Polyclonal
Unconjugated
SEPHS1
1110046B24Rik; AA589574; AI505014; AW111620; RP11-24J20.2; SELD; selenide; Selenide, water dikinase 1; selenium donor protein 1; Selenophosphate synthase 1; selenophosphate synthetase 1; selenophosphate synthetase 1 +E9; selenophosphate synthetase 1 +E9a; selenophosphate synthetase 1 delta E2; selenophosphate synthetase 1 delta E8; selenophosphate synthetase 1 major; SEPHS1; SPS; SPS1
Rabbit
Antigen affinity chromatography
RUO
109079, 22929
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.67 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P49903, Q8BH69
SEPHS1
Recombinant fragment corresponding to a region within amino acids 164 and 392 of Selenophosphate synthetase 1 (Uniprot ID#P49903)
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers