Learn More
SEMA6C Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547255
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human (rh) Semaphorin 6B and rhSemaphorin 6D is observed, 5% cross-reactivity with rhSemaphorin 6A is observed, and less than 3% cross-reactivity with rhSemaphorin 3A, rhSemaphorin 3B, recombinant mouse (rm) Semaphorin 3C, rmSemaphorin 3E, rmSemaphorin 3F, and rmSemaphorin 7A is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
SEMA6C Polyclonal antibody specifically detects SEMA6C in Mouse samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Specifications
SEMA6C | |
Polyclonal | |
Unconjugated | |
SEMA6C | |
KIAA1869; mKIAA1869; m-Sema Y; m-SemaY; m-SemaY2; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6C; sema Y; SEMA6C; semaphorin 6c; semaphorin-6C; Semaphorin-Y; Semay | |
Goat | |
Antigen affinity chromatography | |
RUO | |
20360 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9WTM3 | |
SEMA6C | |
Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 6C Ala26-Ile602 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |