missing translation for 'onlineSavingsMsg'
Learn More

Semaphorin 6C Rat anti-Mouse, Clone: 256631, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB2108

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13429185

  • £388.08 / 500µg

Estimated Shipment date
from Supplier 26-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Semaphorin 6C Monoclonal antibody MAB2108 Specifically detects Semaphorin 6C in Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

Semaphorin 6C
256631
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SEMA6C
Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 6C Ala26-Ile602 Accession # Q9WTM3
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10500
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunohistochemistry
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Q9WTM3
KIAA1869, m-SemaY, m-SemaY2, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6C, Sema Y, Sema6C, Semaphorin Y, semaphorin-6C, semaphorin-Y, SEMAY
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Semaphorin 6C in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) Semaphorin 6C is observed and no cross-reactivity with rhSemaphorin 3A, 6B, recombinant mouse Semaphorin 3B, 3C, 3E, 3F, 6A, 6D, or 7A is observed.
Mouse
Videos
Documents

For Research Use Only