missing translation for 'onlineSavingsMsg'
Learn More

Semaphorin 6C Mouse anti-Human, Clone: 254506, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB2219

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449275

  • £376.32 / 500µg

Estimated Shipment date
from Supplier 17-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Semaphorin 6C Monoclonal antibody MAB2219 Specifically detects Semaphorin 6C in Human samples. It is validated for Western Blotting
Specifications

Specifications

Semaphorin 6C
254506
Western Blot 1 ug/mL
NP_112175
KIAA1869, m-SemaY, m-SemaY2, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6C, Sema Y, Sema6C, Semaphorin Y, semaphorin-6C, semaphorin-Y, SEMAY
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Semaphorin 6C in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Semaphorin 3A, 3B, 6A, 6B, 6D, 7A, recombinant mouse Semaphorin 3C, 3E, 3F, or 6C is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SEMA6C
Mouse myeloma cell line NS0-derived recombinant human Semaphorin 6C Ala25-Val601 Accession # NP_112175
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10500
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only