missing translation for 'onlineSavingsMsg'
Learn More

Semaphorin 6A Rat anti-Mouse, Clone: 201932, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1615

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13468775

  • £388.08 / 500µg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Semaphorin 6A Monoclonal antibody MAB1615 Specifically detects Semaphorin 6A in Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

Semaphorin 6A
201932
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SEMA6A
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Semaphorin 6A Gly19-Thr649 Accession # O35464
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
57556
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunohistochemistry
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
O35464
KIAA1368HT018, SEMA, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A, Sema VIA, Sema6A, SEMA6A1, SEMA6A-1, semaphorin 6A-1, Semaphorin VIA, semaphorin-6A, Semaphorin-6A-1, SEMAQ, VIA
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Semaphorin 6A in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human (rh) Semaphorin 6A is observed and no cross-reactivity with recombinant mouse Semaphorins 3C, 3E, 3F or 7A, or rhSemaphorins 3A, or 3B is observed.
Mouse
Videos
Documents

For Research Use Only