missing translation for 'onlineSavingsMsg'
Learn More

Semaphorin 6A Mouse anti-Human, Clone: 169203, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1146

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448395

  • £420.32 / 500µg

Estimated Shipment
17-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Semaphorin 6A Monoclonal antibody MAB1146 Specifically detects Semaphorin 6A in Human samples. It is validated for Western Blotting
Specifications

Specifications

Semaphorin 6A
169203
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SEMA6A
Mouse myeloma cell line NS0-derived recombinant human Semaphorin 6A Gly19-Thr649 Accession # Q9H2E6
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
57556
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9H2E6
KIAA1368HT018, SEMA, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A, Sema VIA, Sema6A, SEMA6A1, SEMA6A-1, semaphorin 6A-1, Semaphorin VIA, semaphorin-6A, Semaphorin-6A-1, SEMAQ, VIA
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Semaphorin 6A in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) Semaphorin 6A is observed and no cross-reactivity with recombinant human (rh) Semaphorin 3A, rhSemaphorin 3B, or rmSemaphorin 3F is observed.
Human
Videos
Documents

For Research Use Only