missing translation for 'onlineSavingsMsg'
Learn More

SEMA4G Sheep anti-Human, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Brand:  Invitrogen PA547786

Code : NEW

Additional Details : Weight : 0.00010kg

Product Code. 15508282

  • £301.00 / 100µg

Estimated Shipment date
from Supplier 22-06-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

SEMA4G
Polyclonal
Unconjugated
SEMA4G
Lyophilized
SEMA4G
Mouse myeloma cell line NS0-derived recombit human Semaphorin 4G Val18-Leu675
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
0.2 mg/mL
PBS with 5% trehalose and No Preservative
Q9NTN9
SEMA4G, KIAA1619FLJ20590, MGC102867, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4G, semaphorin-4G
Sheep
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
57715
Videos
Documents