missing translation for 'onlineSavingsMsg'
Learn More

Semaphorin 3E Mouse anti-Human, Clone: 400520, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB3239

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13429965

  • £286.65 / 100µg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Semaphorin 3E Monoclonal antibody MAB3239 Specifically detects Semaphorin 3E in Human samples. It is validated for Western Blotting
Specifications

Specifications

Semaphorin 3E
400520
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SEMA3E
Mouse myeloma cell line NS0-derived recombinant human Semaphorin 3E Thr25-Ser775 (Arg557Ala and Arg560Ala) Accession # O15041
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
9723
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
O15041
(semaphorin) 3E, coll-5, KIAA0331M-SEMAH, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema3E, SEMAH, SEMAHM-SemaK, semaphorin-3E
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Semaphorin 3E in direct ELISAs and Western blots. In Western blots, less than 10% cross-reactivity with recombinant human (rh) Semaphorin 3A, 3B, 6A, 6B, 6C, 6D, 7A, rmSemaphorin 3C, 3E, or 3F is observed.
Human
Videos
Documents

For Research Use Only