Learn More
SEMA7A, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SEMA7A.
Marca: Abnova H00008482-A01.50uL
Detalles adicionales : Weight : 0.00010kg
Descripción
The protein encoded by this gene binds to cell surfaces through a glycosylphosphatidylinositol (GPI) linkage. The encoded glycoprotein is found on activated lymphocytes and erythrocytes. This protein may be involved in immunomodulatory and neuronal processes. Defects in this gene can result in loss of bone mineral density (BMD). Three transcript variants encoding different isoforms have been found for this gene
Sequence: EPHKECPNPKPDKAPLQKVSLAPNSRYYLSCPMESRHATYSWRHKENVEQSCEPGHQSPNCILFIENLTAQQYGHYFCEAQEGSYFREAQHWQLLPEDEspecificaciones
SEMA7A | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant SEMA7A. | |
SEMA7A | |
CD108/CDw108/H-SEMA-K1/H-Sema-L/JMH/MGC126692/MGC126696/SEMAK1/SEMAL | |
Mouse | |
50 μL | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_003612 | |
SEMA7A | |
SEMA7A (NP_003603, 536 a.a. ∼ 633 a.a) partial recombinant protein with GST tag. | |
RUO | |
8482 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |