Learn More
Secretory IgA Monoclonal Antibody (Mc29-12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA181174
Code : LD
Additional Details : Weight : 0.01000kg
Description
This clone recognizes purified human secretory chain and secretory IgA. It shows no reaction against myeloma IgA (native or reduced and alkylated), and is negative against normal human IgM, myeloma IgM, normal human IgG; free kappa or gamma chains, and J-chain. This antibody is recommended for frozen and paraffin embedded sections of intestinal and breast tissue.
Secretory IgA Monoclonal antibody specifically detects Secretory IgA in Human samples. It is validated for ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
Secretory IgA | |
Monoclonal | |
1 mg/mL | |
TBS with <0.1% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
Mc29-12 | |
Unconjugated | |
Human IgA | |
Proprietary Immunogen. | |
200 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |