Learn More
EXOC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531529
Code : PN
Additional Details : Weight : 0.01000kg
Description
Predicted reactivity: Mouse (100%), Zebrafish (90%), Rhesus Monkey (98%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EXOC1 Polyclonal antibody specifically detects EXOC1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin).Specifications
EXOC1 | |
Polyclonal | |
Unconjugated | |
EXOC1 | |
2810407P21Rik; A730011E05Rik; BM-012; BM-102; Exoc1; Exocyst complex component 1; exocyst complex component Sec3; Sec3; SEC3L1; SEC3-like 1; Sec3p | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55763, 69940 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.93 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8R3S6, Q9NV70 | |
EXOC1 | |
Recombinant fragment corresponding to a region within amino acids 687 and 894 of Human SEC3 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |