missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSHZ1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527647
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T. Predicted reactivity: Mouse (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TSHZ1 Polyclonal antibody specifically detects TSHZ1 in Human samples. It is validated for Western Blot.Specifications
TSHZ1 | |
Polyclonal | |
Unconjugated | |
TSHZ1 | |
5730407I04Rik; antigen NY-CO-33; CAA; D18Bwg1409e; Kiaa4206; mKIAA4206; Mtsh1; NY-CO-33; SDCCAG33; Serologically defined colon cancer antigen 3 homolog; serologically defined colon cancer antigen 33; Teashirt homolog 1; teashirt zinc finger family member 1; teashirt zinc finger homeobox 1; teashirt1; TSH1; Tshz1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10194 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q6ZSZ6 | |
TSHZ1 | |
Recombinant fragment corresponding to a region within amino acids 468 and 759 of Human SDCCAG33 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |