missing translation for 'onlineSavingsMsg'
Learn More

SCF/c-kit Ligand Mouse anti-Canine, Feline, Clone: 371907, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB2268

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13429315

  • £286.65 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SCF/c-kit Ligand Monoclonal antibody MAB2268 Specifically detects SCF/c-kit Ligand in Canine, Feline samples. It is validated for Western Blotting
Specifications

Specifications

SCF/c-kit Ligand
371907
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
KITLG
E. coli-derived recombinant canine SCF/c-kit Ligand Lys26-Ala190 Accession # Q06220
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4254
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q06220
c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects canine and feline SCF/c-kit Ligand in direct ELISAs and Western blots. In direct ELISAs and Western blots, 50%-100% cross-reactivity with recombinant human (rh) SCF and recombinant mouse SCF is observed and no cross-reactivity with rhM-CSF is observed.
Canine, Feline
Videos
Documents

For Research Use Only