Learn More
SCF Monoclonal Antibody (13302), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523705
Code : NEW
Additional Details : Weight : 0.00010kg
Description
This antibody detects recombinant human SCF in Western blots. In sandwich ELISAs, no significant cross-reactivity or interference was observed with 36 other cytokines. Reconstitute at 0.5 mg/mL in sterile PBS.
SCF Monoclonal antibody specifically detects SCF in Human samples. It is validated for ELISA, Western Blot.Specifications
SCF | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P21583 | |
KITLG | |
E. coli-derived recombinant human SCF Glu26-Ala189 | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
13302 | |
Unconjugated | |
KITLG | |
710-712; blz; c-Kit ligand; Clo; cloud gray; Con; contrasted; CSF; DCUA; DFNA69; DKFZp686F2250; familial progressive hyperpigmentation 2; FPH2; FPHH; Gb; grizzle-belly; hematopoietic growth factor KL; H-SCF; Kit ligand; Kitl; Kitlg; kit-ligand; KL-1; Mast cell growth factor; MGF; M-SCF; Processed kit ligand; SCF; SF; SHEP7; sKITLG; Sl; SLF; Soluble KIT ligand; Steel factor; steel factor/kit ligand; stem cell factor; stem cell factor KL-1; stem cell factor, CSF | |
Mouse | |
Protein A/G | |
RUO | |
4254 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |