missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SATB1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519148
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
SATB1 Polyclonal antibody specifically detects SATB1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
SATB1 | |
Polyclonal | |
Unconjugated | |
Satb1 | |
2610306G12Rik; AW413156; DNA-binding protein SATB1; SATB homeobox 1; Satb1; special AT-rich sequence binding protein 1; special AT-rich sequence binding protein 1 (binds to nuclear matrix/scaffold-associating DNA); special AT-rich sequence-binding protein 1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
6304 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q01826 | |
Satb1 | |
Synthetic peptide sequence (ESDEENRQKTRPRT) corresponding to the internal amino acids of SATB1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |